missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzylamine, 97%, Thermo Scientific™

Product Code. 11468937
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175205-64-8
Molecular Formula C8H9F3NO
Molecular Weight (g/mol) 192.16
MDL Number MFCD00085176
InChI Key FYXMMFFZMQMXCQ-UHFFFAOYSA-O
Synonym 2-trifluoromethoxy benzylamine, 2-trifluoromethoxy phenyl methanamine, 2-trifluoromethoxybenzylamine, o-trifluoromethoxybenzyl amine, 1-2-trifluoromethoxy phenyl methanamine, 2-trifluormethoxybenzylamine, benzenemethanamine, 2-trifluoromethoxy, 2-trifluoromethoxy phenyl methylamine, pubchem4432
PubChem CID 2777251
IUPAC Name [2-(trifluoromethoxy)phenyl]methanamine
SMILES [NH3+]CC1=CC=CC=C1OC(F)(F)F

Specifications

Boiling Point 77°C (12 mmHg)
Refractive Index 1.4531
Quantity 1 g
UN Number UN2735
Beilstein 9257237
Sensitivity Air Sensitive
Formula Weight 191.15
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethoxy)benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'