missing translation for 'onlineSavingsMsg'
Learn More

2'-(Trifluoromethyl)acetanilide, 98%, Thermo Scientific™

Product Code. 11447927
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-62-7
Molecular Formula C9H8F3NO
Molecular Weight (g/mol) 203.164
MDL Number MFCD00013555
InChI Key OXDTZGRSCDEKGO-UHFFFAOYSA-N
Synonym n-2-trifluoromethyl phenyl acetamide, 2'-trifluoromethyl acetanilide, acetanilide, 2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-acetotoluidide, 2-acetamidobenzotrifluoride, n1-2-trifluoromethyl phenyl acetamide, acetamide, n-2-trifluoromethyl phenyl, o-acetaminobenzotrifluoride, o-cf3 acetanilide
PubChem CID 67655
IUPAC Name N-[2-(trifluoromethyl)phenyl]acetamide
SMILES CC(=O)NC1=CC=CC=C1C(F)(F)F

Specifications

Melting Point 94°C to 97°C
Odor Odorless
Quantity 25 g
Beilstein 2106815
Formula Weight 203.16
Percent Purity 98%
Chemical Name or Material 2'-(Trifluoromethyl)acetanilide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'