missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyrrolidine, 97%, Thermo Scientific Chemicals

Product Code. 15284398
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109074-67-1
Molecular Formula C5H8F3N
Molecular Weight (g/mol) 139.12
InChI Key VINAMCOZNJHNIH-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyrrolidine, 2-trifluoromethyl-pyrrolidine, dl-2-trifluoromethylpyrrolidine, pyrrolidine,2-trifluoromethyl, pyrrolidine, 2-trifluoromethyl, +/--2-trifluoromethyl pyrrolidine, acmc-20dptq, acmc-1cih7, 2-trifluoromethyl tetrahydropyrrole
PubChem CID 2782839
IUPAC Name 2-(trifluoromethyl)pyrrolidine
SMILES C1CC(NC1)C(F)(F)F

Specifications

Density 1.2100g/mL
Boiling Point 110.0°C to 112.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.21
Formula Weight 139.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)pyrrolidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(Trifluoromethyl)pyrrolidine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'