missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)ethanol, 99%, Thermo Scientific Chemicals

Product Code. 10512364
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2916-68-9
Molecular Formula C5H14OSi
Molecular Weight (g/mol) 118.25
MDL Number MFCD00002825
InChI Key ZNGINKJHQQQORD-UHFFFAOYSA-N
Synonym 2-trimethylsilyl ethanol, ethanol, 2-trimethylsilyl, 2-hydroxyethyl trimethylsilane, 2-trimethylsilyl ethan-1-ol, silane, 2-hydroxyethyl trimethyl, 2-trimethylsilanyl-ethanol, 2-trimethylsilyl-ethanol, 2-trimethylsilyl ethanal
PubChem CID 18013
IUPAC Name 2-trimethylsilylethanol
SMILES C[Si](C)(C)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.8250g/mL
Boiling Point 71.0°C to 73.0°C (35.0 mmHg)
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)3SiCH2CH2OH
Refractive Index 1.4235 to 1.4255
Quantity 50 mL
Specific Gravity 0.825
Solubility Information Solubility in water: soluble in water
Formula Weight 118.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilyl)ethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(Trimethylsilyl)ethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.