missing translation for 'onlineSavingsMsg'
Learn More

2-[(Trimethylsilyl)ethynyl]aniline, 97%, Thermo Scientific Chemicals

Product Code. 15492328
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103529-16-4
Molecular Formula C11H15NSi
Molecular Weight (g/mol) 189.333
MDL Number MFCD02093646
InChI Key ISXSBRZDPKQYSC-UHFFFAOYSA-N
Synonym 2-trimethylsilyl ethynyl aniline, 2-2-trimethylsilylethynyl aniline, 2-trimethylsilylethynyl aniline, 2-2-trimethylsilyl ethynyl aniline, benzenamine, 2-trimethylsilyl ethynyl
PubChem CID 4377953
IUPAC Name 2-(2-trimethylsilylethynyl)aniline
SMILES C[Si](C)(C)C#CC1=CC=CC=C1N

Specifications

Color Colorless to Yellow
Density 0.94 g/mL
Boiling Point 120°C (0.1 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.564
Quantity 25 g
Formula Weight 189.33
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-[(Trimethylsilyl)ethynyl]aniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'