Learn More
2-(Trimethylsilyl)thiazole, 97%, Thermo Scientific Chemicals
£29.40 - £454.00
Chemical Identifiers
CAS | 79265-30-8 |
---|---|
Molecular Formula | C6H11NSSi |
Molecular Weight (g/mol) | 157.31 |
MDL Number | MFCD00066274 |
InChI Key | VJCHUDDPWPQOLH-UHFFFAOYSA-N |
Synonym | 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq |
PubChem CID | 588453 |
IUPAC Name | trimethyl(1,3-thiazol-2-yl)silane |
SMILES | C[Si](C)(C)C1=NC=CS1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11434977
|
Thermo Scientific Alfa Aesar
B21903.03 |
1 g | N/A | ||||||
11444977
|
Thermo Scientific Alfa Aesar
B21903.06 |
5 g |
£114.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454977
|
Thermo Scientific Alfa Aesar
B21903.14 |
25 g |
£454.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79265-30-8 | |
157.31 | |
VJCHUDDPWPQOLH-UHFFFAOYSA-N | |
588453 | |
C[Si](C)(C)C1=NC=CS1 |
C6H11NSSi | |
MFCD00066274 | |
2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq | |
trimethyl(1,3-thiazol-2-yl)silane |
Specifications
79265-30-8 | |
53°C to 55°C (10 mmHg) | |
C6H11NSSi | |
MFCD00066274 | |
Moisture sensitive | |
VJCHUDDPWPQOLH-UHFFFAOYSA-N | |
trimethyl(1,3-thiazol-2-yl)silane | |
588453 | |
97% |
0.985 | |
57°C (135°F) | |
1.497 | |
UN1993 | |
2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq | |
C[Si](C)(C)C1=NC=CS1 | |
157.31 | |
157.31 | |
2-(Trimethylsilyl)thiazole |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P280-P370+P378q-P501c
H227
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only