missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Undecanol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Melting Point | 1°C to 3°C |
Density | 0.828 |
Boiling Point | 228°C to 229°C |
Flash Point | 107°C (225°F) |
Odor | Mild |
Refractive Index | 1.437 |
Quantity | 10 g |
Beilstein | 1719795 |
Solubility Information | Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether. |
Formula Weight | 172.31 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'