Learn More
2-Undecanone, 98%, Thermo Scientific Chemicals
£18.80 - £133.00
Chemical Identifiers
CAS | 112-12-9 |
---|---|
Molecular Formula | C11H22O |
Molecular Weight (g/mol) | 170.30 |
MDL Number | MFCD00009583 |
InChI Key | KYWIYKKSMDLRDC-UHFFFAOYSA-N |
Synonym | 2-undecanone, methyl nonyl ketone, 2-hendecanone, ketone, methyl nonyl, undecanone, nonyl methyl ketone, rue ketone, 2-oxoundecane, methyl n-nonyl ketone, methylnonylketone |
PubChem CID | 8163 |
ChEBI | CHEBI:17700 |
IUPAC Name | undecan-2-one |
SMILES | CCCCCCCCCC(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10225050
|
Thermo Scientific Acros
140690050 |
5 mL | Glass bottle |
£18.80
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10061630
|
Thermo Scientific Acros
140692500 |
250 mL | Glass bottle |
£41.70
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10093013
|
Thermo Scientific Acros
140690010 |
1 L | Glass bottle |
£133.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-12-9 | |
170.30 | |
KYWIYKKSMDLRDC-UHFFFAOYSA-N | |
8163 | |
undecan-2-one |
C11H22O | |
MFCD00009583 | |
2-undecanone, methyl nonyl ketone, 2-hendecanone, ketone, methyl nonyl, undecanone, nonyl methyl ketone, rue ketone, 2-oxoundecane, methyl n-nonyl ketone, methylnonylketone | |
CHEBI:17700 | |
CCCCCCCCCC(C)=O |
Specifications
112-12-9 | |
100.0 | |
Colorless to Yellow | |
231.0°C to 232.0°C | |
Authentic | |
C11H22O | |
1.4280 to 1.4310 | |
01, 713 | |
0.824 | |
Solubility in water: insoluble | |
CCCCCCCCCC(C)=O | |
170.30 | |
CHEBI:17700 | |
98% | |
2-Undecanone, 99% |
97.5 | |
11.0°C to 13.0°C | |
0.8240g/mL | |
88°C | |
97.5% min. (GC) | |
CH3(CH2)8COCH3 | |
MFCD00009583 | |
15, 6177 | |
2-undecanone, methyl nonyl ketone, 2-hendecanone, ketone, methyl nonyl, undecanone, nonyl methyl ketone, rue ketone, 2-oxoundecane, methyl n-nonyl ketone, methylnonylketone | |
KYWIYKKSMDLRDC-UHFFFAOYSA-N | |
undecan-2-one | |
8163 | |
170.3 | |
Liquid |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-937-5
missing translation for 'rtecsNumber' : YQ2820000
missing translation for 'tsca' : TSCA
- 2-Undecanone
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H410-Very toxic to aquatic life with long lasting effects.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
RUO – Research Use Only