Learn More
Fisherbrand™ Two-Way Scaffolding Connectors
Used for fixing two rods or tubes at right angles. Fisherbrand™ Two-Way Scaffolding Connectors can be tightened by hand.
Brand:  Fisherbrand™ 2114
Product Code. 11518112
  • £18.60 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With thumbscrews
  • For rods and tubes of 6 to 13mm diameters
Specifications
Specifications
2-Way Connector
2-Way
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products