Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ R-A6x250/8MI Fixed Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-A6x250/8MI Fixed Rotors
Brand:  Ohaus™ 30304370
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15896642
  • £189.00 / Pack of 2
In Stock
to see stock.
Description
Description
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-A6x250/8MI Fixed Rotors
Specifications
Specifications
RB
16 mm
Adapter, 5x10ml D16mm RB, 2/pk
2
5 x 10 mL
R-A6x250/8MI Fixed Rotors
5