missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ Rotors

Facilitate centrifugation of tubes smaller than the original, provided carrier space for an array of Frontier™ fixed angle rotors.

Brand:  Ohaus™ 30130891

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15806642

  • £105.00 / Pack of 2
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Accommodates centrifuge tubes that are smaller than the space provided in the rotor carrier.
  • Suitable for use in the following Frontier™ rotors:
    • R-A12x15/6M
    • R-A6x50/6MI
    • R-A12x15/6MI
    • R-A30x15/4MI
    • R-A10x50/10MI
Specifications

Specifications

30 mL
Frontier Fixed-Angle Rotors
1
26 mm
Adapter, D26mm, 30ml Tubes x2
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers