missing translation for 'onlineSavingsMsg'
Learn More

20-Deoxyingenol, MedChemExpress

MedChemExpress 20-Deoxyingenol, a diterpene, is isolated from the roots of Euphorbia kansui. 20-Deoxyingenol can promote autophagy and lysosomal biogenesis by promoting the nuclear translocation of transcription factor EB (TFEB) in vitro. 20-Deoxyingenol can be used for the research of osteoarthritis (OA).

£136.73 - £246.84

Specifications

Chemical Name or Material 20-Deoxyingenol
CAS 54706-99-9
Color White
Purity Grade Notes Research
Molecular Formula C20H28O4
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16469148
Encompass_Preferred
View Documents
Medchem Express
HY-N0866
10 mg
£246.84
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16421558
Encompass_Preferred
View Documents
Medchem Express
HY-N0866
5 mg
£136.73
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

20-Deoxyingenol, a diterpene, is isolated from the roots of Euphorbia kansui. 20-Deoxyingenol can promote autophagy and lysosomal biogenesis by promoting the nuclear translocation of transcription factor EB (TFEB) in vitro. 20-Deoxyingenol can be used for the research of osteoarthritis (OA).
Specifications

Specifications

20-Deoxyingenol
White
C20H28O4
O=C1[C@@]2(C=C(C)[C@@H]3O)[C@]3(O)[C@H](O)C(C)=C[C@@]1([H])[C@@]4([H])[C@@](C4(C)C)([H])C[C@H]2C
332.43
Research
54706-99-9
Research
DMSO : 50 mg/mL (150.41 mM; Need ultrasonic)
332.43
98.0%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.