missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ 20 mL Opus™ Titration Unit
A titration unit with a reagent recirculation system for motor-powered titration directly from the reagent bottle.
Brand: Hirschmann™ 9582020
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Electronic control and dispensing of volumes
- A light protection window for the valve block is also included
- Titration impulse triggered by the motor enables the achievement of a droplet break up from 10 μL for precision determination
- Media recirculating system for simple and rapid venting without reagent loss
- Free rotation
- TFT touchscreen with user-friendly menu
- Method memory for nine different titration procedures with storage of all settings
- USB and RS232 interface for PC connection
Specifications
0.2% | |
50 mL | |
Reagent Circulation | |
Titration Apparatus |