Learn More
Restek™ 20 mm Pressure Release Aluminum Seals with Septa, Assembled
Preassembled seals save time and increase productivity
Brand: Restek™ 21765
This item is not returnable.
View return policy
Description
- Color: Silver
- Material: PTFE/Gray Butyl Rubber Septa
Specifications
Silver | |
PTFE, Butyl Rubber | |
Crimp Cap |
Aluminum | |
100 | |
20 mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Restek™ 20 mm Pressure Release Aluminum Seals with Septa, Assembled
Spot an opportunity for improvement?Share a Content Correction