missing translation for 'onlineSavingsMsg'
Learn More
Learn More
20 NTU Ratio Turbidity Standard, Reagecon
Shop All Reagecon Products

Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
Description
- Turbidity Reference Standard, Turbidity Reference Material.
- Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
- Certified
- NIST traceable
- Non-toxic, non-carcinogenic
- Highly accurate
- Stable
- EPA approved
Specifications
Specifications
| For Use With (Equipment) | Ratio Instruments |
| Description | Liquid format in HDPE Bottle GL45 |
| Accuracy | Accuracy ± 1% NTU |
| Turbidity | 20 NTU ± 1% |
| Volume (Metric) | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction