Learn More
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
Brand:  RETSCH 60.123.045000
 View more versions of this product
Product Code. 10127221
  • £163.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
Test Sieve
Stainless Steel
25 mm
200 mm
Stainless Steel
45 mm
ASTM E 11
Shaker AS 300
For separation, fractioning, particle size determination
Stainless Steel