missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
Brand: RETSCH 60.123.000500
Additional Details : Weight : 0.20000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1Each | |
Test Sieve | |
Stainless Steel | |
25mm |
For separation, fractioning, particle size determination | |
200 mm | |
500μm | |
ASTM E 11 | |
Shaker AS 300 |