Learn More
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
Brand:  RETSCH 60.123.001000
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 10660272
  • £110.00 / Each
Estimated Shipment: 21-10-2024
to see stock.
Add to basket
Specifications
Stainless Steel
Stainless Steel
1Each
Test Sieve
Stainless Steel
25mm
For separation, fractioning, particle size determination
200 mm
1mm
ASTM E 11
Shaker AS 300