Learn More
Eppendorf™ 2000μL Sterile Polypropylene 96-Well Deep Well PCR Plates
GREENER_CHOICEBIOPHARMA
PCR plates, free of DNA, DNase, RNase and PCR inhibitors
Brand:  Eppendorf™ 0030502345
210.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10227512
  • £221.88 / Pack of 20
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
These plates combine a new well design with a sophisticated color coding system for ease of reading and orientation. The well geometry allows high speed pipetting without splashing, excellent mixing and high speed centrifugation, which accelerates your manual and automated work processes. The new polypropylene plates fulfill all current SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines.
  • Recovermax - concaved round bottom well shape with smoothed edges
  • Optitrack - color framed plates with alphanumeric labelling
  • High chemical resistance
  • G-safe - highest centrifugation stability
  • Free from DNA, DNase, RNase and PCR* inhibitor (PCR* clean)
  • Autoclavable
  • Optionally available with barcode
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
20 Plates (5 Bags x 4 Plates
Polypropylene
Sterile
8 x 12
Blue
96
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ 2000μL Sterile Polypropylene 96-Well Deep Well PCR Plates > Color: Blue