missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 2000μL Sterile Polypropylene 96-Well Deep Well PCR Plates
GREENER_CHOICEBIOPHARMA

Product Code. 10267462 Shop All Eppendorf Products
Click to view available options
Color:
Blue
Green
White
Yellow
Packaging:
10 Bags x 8 Plates
5 Bags x 4 Plates
Unit Size:
Pack of 20
Pack of 80
This item is not returnable. View return policy

Product Code. 10267462

Brand: Eppendorf™ 0030502302

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

PCR plates, free of DNA, DNase, RNase and PCR inhibitors

These plates combine a new well design with a sophisticated color coding system for ease of reading and orientation. The well geometry allows high speed pipetting without splashing, excellent mixing and high speed centrifugation, which accelerates your manual and automated work processes. The new polypropylene plates fulfill all current SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines.
  • Recovermax - concaved round bottom well shape with smoothed edges
  • Optitrack - color framed plates with alphanumeric labelling
  • High chemical resistance
  • G-safe - highest centrifugation stability
  • Free from DNA, DNase, RNase and PCR* inhibitor (PCR* clean)
  • Autoclavable
  • Optionally available with barcode
TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 4 Plates
Color White
Product Type Deepwell Plate
Material Polypropylene
Volume (Metric) Well 2000 μL
No. of Wells 96
Sterility Sterile
Well Shape Conical Bottom (V-well)
For Use With (Application) Sample storage and preparation, bacteria and yeast cultivation, DNA and RNA isolation with high throughput
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.