Learn More
Fisher Chemical 200LT ACETONE - CERTIFIED REAGENTCertiFied AR
Brand: Fisher Chemical A/0600/27
1044.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
200LT ACETONE - CERTIFIED REAGENTCertiFied AR
This item is not returnable.
View return policy
Description
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Product Title
Fisher Chemical 200LT ACETONE - CERTIFIED REAGENTCertiFied AR >
Spot an opportunity for improvement?Share a Content Correction