missing translation for 'onlineSavingsMsg'
Learn More

200LT ACETONE - SPECIFIED REAGENT

Brand:  Fisher Chemical A/0560/27

1244.58 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.



200LT ACETONE - SPECIFIED REAGENT

Code : 50

Additional Details : CAS Number : 67-64-1 Weight : 158.00000kg Transport : UN number : 1090 Chem class : 3 Pack group : II


Product Code. 10662241

  • £1315.00 / 200 L
In Stock
to see stock.

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Acetone

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents

Documents

Certificates
Special Offers

Special Offers