Learn More
200LT DICHLOROMETHANE - CERTIFIED REAGEN
Brand: Fisher Chemical D/1852/27
679.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
200LT DICHLOROMETHANE - CERTIFIED REAGEN
Code : 27
Additional Details : CAS Number : 75-09-2 Weight : 265.00000kg Transport : UN number : 1593 Chem class : 6.1 Pack group : III
Safety and Handling
- Dichloromethane
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.