Learn More
200LT PROPAN-2-OL - SPECIFIED REAGENT
Brand:  Fisher Chemical P/7490/27
200LT PROPAN-2-OL - SPECIFIED REAGENT
Code : 27
Additional Details : CAS Number : 67-63-0 Weight : 157.00000kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Product Code. 11357670
  • £1805.00 / 200 L
Estimated Shipment: 05-09-2024
to see stock.
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Isopropanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
SDS Documents