Learn More
RETSCH 203.2 dia. x 50.8mmH Stainless Steel Test Sieve, ISO Certificate, Micrometer Pore Sizes
Brand: RETSCH 60.149.000450
This item is not returnable.
View return policy
Specifications
1Each | |
203.2 mm | |
Stainless Steel | |
Test Sieve | |
Stainless Steel | |
50.8mm |
For separation, fractioning, particle size determination | |
Stainless Steel | |
450 μm | |
ISO 3310/1 | |
Shaker AS 300 |