Learn More
205LT ETHANOL BP EP USP
Brand: Fisher Chemical E/0650DF/28
2784.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
205LT ETHANOL BP EP USP
Code : 27
Additional Details : CAS Number : 64-17-5 Weight : 161.95000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Safety and Handling
- Ethanol 96%, technical, duty free
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.