missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- 20mm
- aluminum Cap
- Gold
- Center hole
- Combination Seal
Specifications
Butyl | |
20 mm | |
Headspace | |
3 mm |
Dark Gray, Gold Lacquered | |
Aluminum | |
100 | |
Crimp Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction