missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Beta Catenin (Ser675) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28853-1-AP-20UL
This item is not returnable.
View return policy
Description
Phospho-Beta Catenin (Ser675) Polyclonal antibody specifically detects Phospho-Beta Catenin (Ser675) in Human, Mouse samples. It is validated for WB, ELISASpecifications
Phospho-Beta Catenin (Ser675) | |
Polyclonal | |
Unconjugated | |
BC014566 | |
Protein A purification | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot, ELISA | |
450 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
Rabbit | |
20 μL | |
1499 | |
Store at -20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction