Learn More
Tuttnauer 21 CFR Part 11 Software
Product Code. 17829558 Shop All Tuttnauer Products
Click to view available options
:
21 CFR Part 11 Software
Product Code. 17829558
Brand: Tuttnauer ADD2220442
Only null left
Add to basket
Add to basket
Record of performed cycles and of actions taken by operator using software and system for operating the autoclave. R.PC.R is required for 21 CFR part 11 in order to save a history log of cycles and track changes made to the autoclave parameters by operators.
  • Record of performed cycles.
  • Record of actions taken by operator using software and system for operating the autoclave (according to user access levels).
  • This information can be used for auditing the operation of the autoclave.
  • R.PC.R is required for 21 CFR part 11 in order to save a history log of cycles and track changes made to the autoclave parameters by operators.
  • 21 CFR part 11 - Determined by the manufacturers policy.
Specifications
Certifications/Compliance 21 CFR Part 11
Product Type Software