Learn More
uvex 2112 One-Fit Disposable Ear Plugs
Characterized by their ergonomic, closed surface design One-Fit ear plugs are constructed of polyurethane foam for optimal comfort, even during long periods of use.
Brand:  uvex 2112046
24.63 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17200342
  • £34.00 / 200 pairs
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Soft, polyurethane foam ensures comfort even during long periods of use
  • Closed surface prevents foreign bodies from entering
  • For use with noise sources up to 110 db(A)
  • Cordless, ergonomic design
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turquoise
Regular
Refill Bag
37 dB
Oval
Single-use
No
No
2112 One-Fit
200
Uncorded
Ear Plugs
No
EN 352-2