Learn More
Implen 21CFR Part 11 Software for the NanoPhotometer with IQ/OQ package
Software upgrade and IQ/OQ package for NanoPhotometer models for compliance with 21 CFR part 11 FDA guidelines.
£3690.00 - £4150.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17720248
|
Implen
CFR21-Q-120 |
NanoPhotometer N120 |
£3690.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17730248
|
Implen
CFR21-Q-60 |
NanoPhotometer N60 |
£3690.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17740248
|
Implen
CFR21-QHN-40 |
NanoPhotometer C40 |
£4150.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17750248
|
Implen
CFR21-QHN-80 |
NanoPhotometer NP80 |
£4150.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- License key provided on USB flash drive
- IQ/OQ documentation provided as Excel file on USB flash drive
- Incl. standard solution N-120-S, N-568-S and or Holmium Glass Filter and Neutral Density Glass Filter Set N-4080-HNF according to NanoPhotometer model
- Software is available for use on touchscreen and computers
- Authentication of user login information
- Ability to set Four Eye Administrator Rights, password guidelines and expiry dates
- Lockout access and re-authentication requirements after a period of inactivity
- System lockout upon unauthorized attempts to access software
- Individual Role Based Access Control for Administrator, Group, Power User and User roles
- Prevention of data tampering
- Generation of searchable audit trail log in human-readable format
- Tracking of changes and committed actions to user roles, account management, and system settings
- Ability to sign records electronically
Specifications
Software | |
IQ/OQ Package |
For research only