Learn More
Tenak™ 22 mm Cryobox Dividers
Dividers for use with 32 mm high cryoboxes.
£37.62 - £45.45
Specifications
Type | Grid Divider |
---|---|
For Use With (Equipment) | 32 mm high cryoboxes |
Height (Metric) | 22 mm |
Product Code | Brand | Tube Array | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tube Array | Holds | Price | Quantity & Availability | ||||
15200624
|
Tenak™
TE22675 |
8 x 8 cells | Vials with 15.50 mm O.D. |
£41.55
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
15210624
|
Tenak™
TE22676 |
7 x 7 cells | Vials with 17.75 mm O.D. |
£41.55
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
15220534
|
Tenak™
TE22092 |
9 x 9 cells | Vials with 13 mm O.D. |
£37.65
Pack of 36 |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
15230534
|
Tenak™
TE22094 |
12 x 12 cells | Vials with 10 mm O.D. |
£41.55
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
15240534
|
Tenak™
TE22093 |
10 x 10 cells | Vials with 12 mm O.D. |
£37.65
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
15240614
|
Tenak™
TE22669 |
16 x 16 cells | Vials with 7.50 mm O.D. |
£45.45
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
15250534
|
Tenak™
TE22095 |
13 x 13 cells | Vials with 9 mm O.D. |
£41.55
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
15250614
|
Tenak™
TE22670 |
14 x 14 cells | Vials with 8.75 mm O.D. |
£45.45
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
15260614
|
Tenak™
TE22671 |
13 x 13 cells | Vials with 9.50 mm O.D. |
£45.45
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
15270614
|
Tenak™
TE22672 |
12 x 12 cells | Vials with 10.25 mm O.D. |
£45.45
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
11581665
|
Tenak™
TE22096 |
14 x 14 cells | Vials with 8 mm O.D. |
£41.55
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
11591665
|
Tenak™
TE22097 |
16 x 16 cells | Vials with 7 mm O.D. |
£41.55
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
15200534
|
Tenak™
TE22090 |
7 x 7 cells | Vials with 17 mm O.D. |
£37.62
Pack of 36 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
Description
- For use in cardboard cryoboxes
- Holds cryogenic vials
- Grid style divider
Specifications
Grid Divider | |
32 mm high cryoboxes | |
22 mm |