missing translation for 'onlineSavingsMsg'
Learn More

Moldex™ 2360 Series P100 Particulate Respirator with Ventex™ Valve

P100 particulate respirator with Ventex exhalation valve allows hot air to escape faster to give wearer a cooler and easier breathing experience.

£67.65

Specifications

Aerosol Type P - Oil-Proof
ANSI Standard ANSI/ISEA 110-2003, Section 7.11.1
Certifications (NIOSH) NIOSH 42 CFR 84, P100
Color White
Design Type Cup/Molded
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12362289
View Documents
Moldex™
236015
1 Ea.
£67.65
Pack of 20
In Stock
to see stock.
Add to basket
 
Description

Description

Application

  • Use in environments where an OSHA substance-specific standard applies (lead, cadmium, arsenic, MDA) as well as pharmaceutical manufacturing, conventional and nuclear power generation, welding, cutting and metal pouring
  • Design
    • Adjustable cloth straps assure snug, comfortable fit
    • Ventex valve lets the hot air out faster so workers breathe easier and stay cooler
    • Soft foam face cushion for added comfort and no pressure points
    • Dura-Mesh™ shell resists collapsing in regular use and even under hot and humid conditions
    • Contour-molded nose bridge seals without metal noseband
    • Single use

    Do not use against concentrations of particulates that exceed the maximum use concentration or 10X the OSHA PEL, whichever is lower.

    Specifications

    Specifications

    P - Oil-Proof
    NIOSH 42 CFR 84, P100
    Cup/Molded
    P100
    General Purpose Particulate Respirator
    Yes
    Environments where an OSHA substance specific standard applies (lead, cadmium, arsenic, mda) as well as pharmaceutical manufacturing, conventional and nuclear power generation, welding, cutting and metal pouring
    Adjustable Cloth Straps
    ANSI/ISEA 110-2003, Section 7.11.1
    White
    Single-use
    Dura-Mesh
    Medium/Large
    99%
    Molded Nose Bridge
    No
    Videos
    SDS
    Documents

    Documents

    Product Certifications