missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3500xL Genetic Analyzer 24-Capillary Array, 50 cm

Use for fragment analysis and rapid sequencing applications.

Brand:  Applied Biosystems™ 4404689

Product Code. 15819722

  • £2075.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

The internally uncoated capillaries are supplied in preassembled sets of 24 capillaries per array, with a built-in frame for easy installation.

Order Info

Each

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fragment Analysis
Store this array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to the Applied Biosystems 3500/3500xL Genetic Analyzer User Guide.
Protein Quality Analysis, Fragment Analysis, Sequencing
50 cm (Capillary)
1 array
Room Temperature
50 cm
24-Capillary Array, 50cm Capillary, 125 min. Run Time, Internally Uncoated Capillaries, Fragment Analysis, Protein Quality Analysis, Sequencing Applications, For Use With 3500xL Genetic Analyzer, Room Temperature
3500xL Genetic Analyzer
Genetic Analyzer 24-Capillary Array
125 min
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM519L67 Certificate TypeCertificate of Analysis Date17/03/2020 Catalog Number4404689
Lot NumberM519G69 Certificate TypeCertificate of Analysis Date05/11/2019 Catalog Number4404689
Lot NumberM519F77 Certificate TypeCertificate of Analysis Date08/10/2019 Catalog Number4404689
Lot NumberM519G55 Certificate TypeCertificate of Analysis Date08/10/2019 Catalog Number4404689
Lot NumberM517D26 Certificate TypeCertificate of Analysis Date17/02/2019 Catalog Number4404689
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ 3500xL Genetic Analyzer 24-Capillary Array, 50 cm > 50cm Capillary Length

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.