missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ 3500xL Genetic Analyzer 24-Capillary Array, 50 cm
Use for fragment analysis and rapid sequencing applications.
Brand: Applied Biosystems™ 4404689
This item is not returnable.
View return policy
Description
The internally uncoated capillaries are supplied in preassembled sets of 24 capillaries per array, with a built-in frame for easy installation.Order Info
Each
Specifications
Fragment Analysis | |
Store this array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to the Applied Biosystems 3500/3500xL Genetic Analyzer User Guide. | |
Protein Quality Analysis, Fragment Analysis, Sequencing | |
50 cm (Capillary) | |
1 array | |
Room Temperature |
50 cm | |
24-Capillary Array, 50cm Capillary, 125 min. Run Time, Internally Uncoated Capillaries, Fragment Analysis, Protein Quality Analysis, Sequencing Applications, For Use With 3500xL Genetic Analyzer, Room Temperature | |
3500xL Genetic Analyzer | |
Genetic Analyzer 24-Capillary Array | |
125 min |
For Research Use Only. Not for use in diagnostic procedures.