Learn More
Thermo Scientific™ 24 Listeria Enrichment Broth (LEB) Buffer Supplement
Optimize growing conditions for all Listeria species prior to molecular assay with Thermo Scientific™ 24 LEB Buffer Supplement, added to our range of complementary media.
£53.40
Specifications
Description 24 LEB Buffer Supplement
Product Type Microbiology Media
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13245549
View Documents Promotion Details
Thermo Scientific™
BO1204M
10/Pk.
£53.40
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
Description
Description
Enrich Listeria monocytogenes and Listeria spp. from food and environmental samples prior to molecular assay with the Thermo Scientific™ 24 Listeria Enrichment Broth (LEB) Buffer Supplement. The supplement is part of the 24 LEB range which includes complementary media and provides optimal growing conditions for all Listeria species for PCR applications.  It is specifically designed for use with PCR Systems, such as Thermo Scientific™ SureTect™ molecular assays for Listeria.
The 24 LEB product range provides rapid growth of all Listeria species and has been validated with Thermo Scientific™ SureTect™ molecular assays for Listeria to provide results in 24 hours from food and environmental samples.
Achieve optimal growing conditions for all Listeria species with 24 LEB Buffer Supplement added to 24 LEB Broth (BO1205S) and 24 LEB Dehydrated Media Complete Base (CM1154).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
24 LEB Buffer Supplement
Microbiology Media
Videos
SDS
Documents
Documents
Product Certifications
Laboratory Use Only