missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Meker Safety Burner for LP Gas, 24mm

Product Code. 12882144 Shop All Fisherbrand Products
Click to view available options
Gas Type:
LPG
Unit Size:
Each

Product Code. 12882144

Only null left
Add to basket
Add to basket

Supplied with a flame failure device that shuts off the gas supply to the burner within approximately 20 seconds of the flame being extinguished.

  • Safety features all contained within the burner
  • For use with natural gas/methane
  • 12.7mm diameter opening

Specifications

Product Type Meker Burner
Gas Type LPG
Diameter (Metric) 24 mm
No. per Pack 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.