missing translation for 'onlineSavingsMsg'
Learn More

Nalgene 24 oz. OTF Bottle

Product Code. 17120066
Click to view available options
Color:
Aqua Blue
Blue
Coral
Gray
This item is not returnable. View return policy
This item is not returnable. View return policy

Bottle with a lock-top and pop-top for one-handed sipping on the fly.

  • Slim shape and fortified cap
  • Ideal for care-free storage in work bags, gym bags, backpacks, and cup holders
  • Opens easily for convenient one-hand refreshment on the fly
  • Locks to prevent spills
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 710 mL
Capacity (English) 24 oz.
Color Aqua Blue
Closure Color Aqua Blue
Closure Type Locking
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'