Learn More
Thermo Scientific™ 24-Place Fixed-Angle Rotors with Covers
£1430.00
Product Code | Brand | Angle | Max. RCF | Max. Speed | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Angle | Max. RCF | Max. Speed | Price | Quantity & Availability | |||
10593851
|
Thermo Scientific™
75007593 |
45° | 21,890 x g | 15,000 rpm |
£1430.00
Each |
Estimated Shipment: 13-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Specifications
Microliter Rotor | |
24 x 1.5/2 mL |