missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ 24-well Insert 2in1, PC-Membrane

The cellGrade™ plus surface of the membranes ensures excellent cell attachment and growth. The highly transparent membranes give you good visibilty of your cell cultures, and the high pore density contributes to good permeability.

Brand:  BRAND™ 782701

249.82 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 16174947

  • £304.00 / Pack of 48
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Use Inserts in a hanging or standing position
  • For 24-well plates or culture dishes
  • Achieve uniform and controlled cell growth due to surface treatment, without coatings
TRUSTED_SUSTAINABILITY
Specifications

Specifications

24-well Insert 2in1, PC-Membrane 0.4 μm, Multipack
0.4 μm
Sterile
Polycarbonate
4 x 12
24
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers