Learn More
BRAND™ 24-well Insert 2in1, PC-Membrane
The cellGrade™ plus surface of the membranes ensures excellent cell attachment and growth. The highly transparent membranes give you good visibilty of your cell cultures, and the high pore density contributes to good permeability.
Brand: BRAND™ 782706
276.73 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
- Use Inserts in a hanging or standing position
- For 24-well plates or culture dishes
- Achieve uniform and controlled cell growth due to surface treatment, without coatings
Specifications
24-well Insert 2in1, PC-Membrane 8 μm | |
8 μm | |
Sterile |
Polycarbonate | |
48 x 1 | |
24 |