Learn More
MP Biomedicals™ QuickPrep™ 2 Sample Holder
Compatible with the QuickPrep System. QuickPrep™ 2 Sample Holder is a 24 x 2 mL QuickPrep sample tube holder for room temperature lysis and grinding on FastPrep-24.
Brand: MP Biomedicals™ 116005512
376.15 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 2.00000kg
Description
- Adapter for FastPrep-24 Instruments to grind, lyse, homogenize 24 x 2 mL samples.
- The QuickPrep adapter is ideal for high-throughput applications and can process up to 24 x 2 mL samples simultaneously.
- It is the standard adapter supplied with the FastPrep-24™ 5G benchtop instrument.
- The ergonomic design of the adapter ensures the 2 mL tubes are easy-to-load and they remain securely sealed during grinding, lysing and homogenization.
- It is stable and can be placed on a workspace to be used as a tube rack.
- Frozen samples loaded in the adapter and stored at -20°C to -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
- Purchase multiple adapters to use the same FastPrep-24 5G instrument for multiple lab groups or sample applications.
Specifications
24 x 2 mL | |
1 Ea. | |
Tube Holder |
Room Temperature (15°C to 30°C) | |
FastPrep-24 |