missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 24mm PP Screw Seal, White, Closed Top, 24-400 Thread, Assembled Septum
36.84 GBP valid until 2024-12-31
Use promo code "21972" to get your promotional price.
Description
- No time consuming and tricky assembly
- No contamination of the liner with sweat/fat normally caused by manual assembly
- Broad variety of different septa materials for almost all applications
Specifications
PTFE/EPDM | |
24 mm | |
Storage | |
2 mm |
Black, White | |
Polypropylene | |
100 | |
Screw Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction