missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ 25 mL Class B Amber Lateral Glass Stopcock Burette

For volumetric accuracy in liquid handling.

Brand:  Hirschmann™ 3220170

Product Code. 13127999

  • £150.00 / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Class B
  • Manufactured from DURAN™ borosilicate glass for resistance to chemicals and thermal shock
  • In amber glass for the protection of light-sensitive contents
  • Lateral glass stopcock
  • Calibrated for EX
  • Acc. to DIN EN ISO 385
  • White graduation marks
Specifications

Specifications

Amber
500 mm
0.05 mL
Lateral
±0.045 mL
Class B
25 mL
Glass
White
Glass
Buret
DIN EN ISO 385
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hirschmann™ 25 mL Class B Amber Lateral Glass Stopcock Burette > Capacity: 25 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.