Learn More
Greiner Bio-One 25 mL Sterile Serological Pipets
GREENER_CHOICE
Product Code. 10449943
Click to view available options
Packaging Type:
Bulk
Individual Plastic Wrap
Unit Size:
Pack of 200
Product Code. 10449943
Brand: Greiner Bio-One 760160
Only null left
Add to basket
Add to basket
Drip-free pipetting design with high optical clarity and maximum accuracy.
  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
TRUSTED_SUSTAINABILITY
Specifications
Description Shape: Standard
Material Polystyrene
Sterility Sterile
Graduated Yes
Graduations 2/10
Packaging Type Individual Plastic Wrap
Volume (Metric) 25 mL
Type Serological Pipet