Learn More
Hanna Instruments™ 25 Tests Reagent Set For Ammonia in Salt Water
Brand: Hanna Instruments™ HI3826025
Code : 04
Additional Details : CAS Number : 7774-29-0 Weight : 0.05000kg Transport : UN number : 2922 Chem class : 8 Pack group : II
Specifications
For Ammonia in Fresh Water | |
25 | |
0.0 to 2.5mg/L | |
Reagent Set |
With Protective Cap, Batteries and Instructions | |
1 Set | |
Nessler Colorimetric |