Learn More
Axygen™ 250 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
GREENER_CHOICE
For use with Agilent/Velocity11 automation systems
Brand:  Axygen™ VT-250-R-S
 View more versions of this product
Product Code. 12585567
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
  • Clear polypropylene
  • Maxymum Recovery™ improves accuracy and precision while reducing sample loss
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Available in nonsterile or sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 μL
Sterile
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Rack
SBS Rack
Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT
Non-filtered
Clear
DNase, Rnase Free
Non-pyrogenic
Automation, Round
50 Racks/Cs.
Polypropylene