Learn More
Code Corporation™ 2700 Palm Barcode Scanner
Read wide and small barcodes easier with this ultra-fast barcode reader. Get unbeatable performance from this aggressive, omnidirectional scanner that reads even damaged barcodes.
Brand: Code Corporation™ 176686
Additional Details : Weight : 0.13100kg Transport : UN number : 3481 Chem class : 9
Description
- Read wide and small barcodes easier with this ultra-fast barcode reader.
- Get unbeatable performance from this aggressive, omnidirectional scanner that reads even damaged barcodes.
- Get your scanning processes started or add this scanner + battery kit to your current scanning solution.
- Uses B27 batteries with built-in power gauge.
- IP65 seal rating against dust and liquid ingress, CodeShield™ housing for disinfectant and chemical wipedown.
- Unequaled performance reading on curved and shiny surfaces.
- USB, Bluetooth™5 Low Energy (for enhanced data security, extended battery life and less downtime).
- Customisable visual, audible and haptic feedback with multiple programmable buttons.
- Trace any troubles – Trackable service dates and battery statistics to help IT deploy and manage their scanner fleet.
Specifications
Code Reader™ 2700 Palm Barcode Scanner | |
Barcode Scanner | |
1 Ea. | |
BC412, Codabar, Code 11, Code 128, Code 32, Code 39, Code 93, GS1 DataBar, Hong Kong 2 of 5, IATA 2 of 5, Interleaved 2 of 5, MSI Plessey, Matrix 2 of 5, NEC 2 of 5, Pharmacode, Plessey, Straight 2 of 5, Telepen, Trioptic, UPC / EAN / JANCodablock F, Code 49, GS1 Composite (CC-A/CC-B/CC-C), MicroPDF, PDF417Aztec Code, Data Matrix, Data Matrix Rectangular Ext'n, GoCode&trade, (prorietary--add'l license req'd), Grid Matrix, HanXin Code, Micro QR Code, QR Code, QR Model 1Australian Post, Canada Post, Itelligent Mail, Japan Post, KIX Code, Korea Post, Planet, Post-Net, UK Royal Mail, UPU ID-tags | |
28.00 mm | |
53.34 mm | |
131.00 mm | |
131 g | |
0.29 lb. | |
Asset tracking, General identification | |
CR2700 Palm Barcode Scanner and B27 Battery | |
3 Years |
Rechargeable Battery | |
None Included | |
Bluetooth | |
Bluetooth 5 Low Energy (Class II) | |
1.1 in. | |
2.1 in. | |
5.2 in. | |
USB 2.0 (Generic HID, HID Keyboard, Virtual COM Port) | |
CE, Europe: CE-RoHS | |
5% to 95% (Non-condensing) | |
Bluetooth 5 Low Energy (Class II) | |
JavaScript |