Learn More
Gibco™ 293fectin™ Transfection Reagent
293fectin™ Transfection Reagent is a proprietary, cationic lipid-based formulation for transfecting DNA into eukaryotic cells.
£598.00 - £63500.00
Specifications
Cell Type | Established Cell Lines |
---|---|
Sample Type | Plasmid DNA |
Transfection Technique | Lipid-based Transfection |
Product Line | 293fectin™ |
Product Type | Transfection Reagent |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10553283
|
Gibco™
12347019 |
1 mL |
£598.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10535675
|
Gibco™
12347500 |
15 mL |
£7160.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10093574
|
Gibco™
12347750 |
10 x 15 mL |
£63500.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
293fectin™ Transfection Reagent is a proprietary, cationic lipid-based formulation for transfecting DNA into eukaryotic cells. This reagent is optimized for transfecting suspension 293 human embryonic kidney cells (e.g., FreeStyle™ 293-F cells, Cat. No. R790-07) in defined, serum-free FreeStyle™ 293 Expression Medium (12338-018), and is intended for use with the FreeStyle™ 293 Expression System (K9000-01).
293fectin™ Transfection Reagent provides the following advantages:
- Demonstrates high transfection efficiency in suspension 293 cells and is also suitable for transfecting adherent 293 cells
- Suspension FreeStyle™ 293-F cells may be transfected in FreeStyle™ 293 Expression Medium; no medium change is required
- Add 293fectin™ reagent complexes directly to cells in culture medium
- No need to remove complexes or change or add medium following transfection
Specifications
Established Cell Lines | |
Lipid-based Transfection | |
Transfection Reagent |
Plasmid DNA | |
293fectin™ | |
Yes |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.