Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors with reducers, injection site
The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.
Marque: Thermo Scientific™ SS00031-I
Informations supplémentaires : Weight : 1.00000kg
Description
The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.
Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions
Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands
BPC port specifications:
• Line 1: Luer lock insert connection with 4.8 mm (3/16 in.) barb, cap, slide clamp, and 4.8 x 4.1 mm (3/16 x 5/32 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
• Line 2: Luer lock body connection with 0.56 cm (0.22 in.) barb, plug, slide clamp, and 0.48 x 0.41 cm (0.12 x 0.16 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
• Line 3: Tubing: EVA, injection port
Spécification
500 mL | |
Store at room temperature. | |
One 4-mil. poly bag | |
Irradiated | |
Labtainer™ | |
ASI 26/77 |
Polyethylene | |
25.9 x 13.7 cm (10.2 x 5.4 in.) | |
50/Cs. | |
Room Temperature | |
Luer Lock Body Connector and Injection Site | |
TPE and EVA |